You Searched For: Dropper Caps
13  results were found
SearchResultCount:"13"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Dropper cap, Thread: 25 mm, For bottles: 250/500 ml
Supplier: DWK Life Sciences
Description: LDPE, coloured.
Supplier: DWK Life Sciences
Description: Screw cap made of radiation grade polypropylene.
Catalog Number: (NALGDS2176-0384)
Supplier: Thermo Fisher Scientific
Description: PP, translucent. with polyethylene insert.
UOM: 1 * 6 items
Catalog Number: (215-2471)
Supplier: Kautex
Description: Combination closure with screw-on dropping tip, Combination closure with screw-on dropping tip, HDPE, Thread: 18 mm, HDPE, For bottles: 10/25/50 ml
UOM: 1 * 1 items
Supplier: Kautex
Description: LDPE, natural colour.
Supplier: BÃœRKLE
Description: Dropper cap with captive cap, for threads Ø 18 mm, Thread: 18 mm, For bottles: 100/250/500/1000 ml
Supplier: KARTELL
Description: HDPE, natural colour.
Sale
Supplier: MUELLER & KREMPEL
Description: Glass pipette mounts for dropping bottles with white PE screw cap and blue rubber hat.
Supplier: MUELLER & KREMPEL
Description: Vertical dropper for aqueous solutions; White tamper evident closure with dropper insert for openings according to DIN-18 for low surface tension aqueous solutions; also containing ethanol up to 15%
Supplier: GLASWARENFABRIK KARL HECHT
Description: Dropper closure for dropping bottles, Screw caps, UNI 1, fast dropping, 4 holes, white, Thread: DIN 18, For bottles: 10/20/30/50/100 ml
Supplier: Brand
Description: Cap with dropper nozzle for dropping bottle, Thread: GL 25, LDPE, For bottles: 250+500 ml
Catalog Number: (HACH2318506)
Supplier: Hach
Description: Dropper cap, with 0,5 and 1,0 ml marks on glass barrel
UOM: 1 * 6 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 13 of 13
no targeter for Bottom