You Searched For: Closures
643  results were found
SearchResultCount:"643"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Vent cap for 125 ml
Supplier: VWR Collection
Description: VWR®, Chromatography vial closure, Crimp caps with septa, open top, Hardness: 50° shore A, Septum: Cream silicone/red PTFE, Thickness: 1,5 mm
Supplier: VWR Collection
Description: VWR®, Chromatography vial closure, PP, Screw caps with septa, open top, 8-425 thread, Hardness: 45° shore A, Septum: Transparent blue silicone/white PTFE, Thickness: 1,3 mm
Supplier: VWR Collection
Description: VWR®, Chromatography vial closure, Snap caps, open top, Hardness: 55° shore A, Septum: White silicone/blue PTFE, Y-slit, Thickness: 1,0 mm
Supplier: VWR Collection
Description: VWR®, Chromatography vial closure, PE, Ø: 11 mm, Type: PE transparent, soft version, Hardness: 60° shore A, Septum: Natural rubber red-orange/TEF transparent, Thickness: 1,0 mm
Sale
Supplier: VWR Collection
Description: VWR®, Chromatography vial closure, Steel, Crimp caps with septa, open top, magnetic, 8 mm hole, Hardness: 50° shore A, Septum: Pharma-fix bromo-butyl/PTFE, Thickness: 3,0 mm
Supplier: VWR Collection
Description: Flat screw cap with O-ring for microcentrifuge tubes, VWR®, Colour: Assorted
Supplier: VWR Collection
Description: VWR®, Chromatography vial closure, Screw caps with septa, open top, EPA quality, Hardness: 45° shore A, Septum: Natural silicone/beige PTFE, bonded, Thickness: 3,2 mm
Supplier: VWR Collection
Description: VWR®, Chromatography vial closure, Screw caps with septa, open top, 10-425 thread, Hardness: 60° shore A, Septum: Red-orange natural rubber/transparent TEF, Thickness: 1,3 mm
Supplier: VWR Collection
Description: VWR®, Chromatography vial closure, PP, Screw caps, open top, Hardness: 55° shore A, Septum: White silicone/blue PTFE, slit, Thickness: 1,0 mm
Supplier: VWR Collection
Description: Chromatography vial closure, Screw caps with septa, open top, Hardness: 45° shore A, Septum: Transparent blue silicone/white PTFE, Thickness: 1,7 mm
Supplier: VWR Collection
Description: Chromatography vial closure, Screw caps with septa, closed top, Hardness: 50° shore A, Septum: Cream silicone/red PTFE, Thickness: 1,5 mm
Supplier: VWR Collection
Description: Safe-T-Flex caps feature a unique, flexible, over-locking design, locking securely on the outside of the tube lip and can be removed with one hand. A ...
Catalog Number: (548-0661A)
Supplier: VWR Collection
Description: VWR®, Chromatography vial closure, Screw cap, closed top, Hardness: 53° shore D, Septum: PTFE virginal, Thickness: 0,5 mm
UOM: 1 * 1.000 items
Supplier: VWR Collection
Description: Crimp Caps ND8 can be purchased with a variety of septa materials. Furthermore, push-on snap caps for crimp neck vials ND8 are available.
Supplier: VWR Collection
Description: VWR®, Chromatography vial closure, PP, Ø: 18 mm, Type: PP black, closed, without septum, Hardness: -, Septum: -, Thickness: - mm
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 643
no targeter for Bottom