Insulating electrode clamp
Supplier: VWR Collection
241-0079EA 137 EUR
VWRF08466.024
Insulating electrode clamp
PMAA, with stainless steel rod.
  • For two electrodes
  • Spanning width up to 20 mm
  • Rod (125×8 mm) can be attached to stand