VWR®, Inoculating Loops and Needles, Disposable
Supplier: VWR
VWR®
612-2495EA 211 EUR
612-2495 612-2496 612-2497 612-2498
VWR®, Inoculating Loops and Needles, Disposable
Spreaders Inoculating Loops and Needles
PS, sterile. Calibrated, disposable inoculating loops made from flexible or rigid plastic that cater for microbiologist's different applications and methods.
  • Colour coded for easy product identification
  • Conveniently packed in anti-roll tubes with push-on resealable cap
  • Fast, easy and secure access to the loops, without the need for a separate holder
  • Loops have an ultra-smooth surface for easy streaking and length of 210 mm
Delivery information: Calibration certificate supplied with every pack.