VWR®, Inoculating Loops and Needles, Disposable
Supplier: VWR
VWR®
612-9358EA 67.6 EUR
612-9358 612-9357 612-9356 612-9355 612-9354 612-9353 612-9352 612-9351
VWR®, Inoculating Loops and Needles, Disposable
Spreaders Inoculating Loops and Needles
PS, sterile. These loops ensure consistent wetting and complete liquid transfer. Available rigid or flexible and have smooth edges to avoid gouging the surface of the media.
  • Gamma sterilised
  • Colour coded
  • Hexagonal shaft for easy handling
Delivery information: Supplied with a calibration certificate in each box.