You Searched For: Carriers
37  results were found
SearchResultCount:"37"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: The safety bottle carrier provides a safe and reliable way to transport a variety of bottled reagents in the lab. Available in 1,5 and 4,5 L capacitie...
New Product
Supplier: VWR Collection
Description: Rubber, coloured.
Supplier: VWR Chemicals
Description: HDPE.
Supplier: VWR Collection
Description: These bottle carriers are made from polyamide coated stainless steel.
Supplier: VWR Collection
Description: Bottle carrier/container, VWR®
Supplier: VWR Collection
Description: Sturdy container for transport of biological samples taken at patient’s home.
Catalog Number: (215-3582)
Supplier: VWR Collection
Description: Glass bottles are designed to contain liquids, but when they break, hazardous liquids potentially mix with broken glass. This bottle carrier protects ...
UOM: 1 * 1 items
Catalog Number: (215-7991)
Supplier: Thermo Fisher Scientific
Description: PC.
UOM: 1 * 1 items
Supplier: BOCHEM
Description: Safety transport container, made of stainless steel, for laboratory bottles.
Catalog Number: (NALG6565-0001)
Supplier: Thermo Fisher Scientific
Description: HDPE, blue.
UOM: 1 * 4 items
Catalog Number: (WILMWG-6192)
Supplier: SP Industries
Description: Made from unbreakable polymer material with a snap-on cap, these tube carriers feature a shirt clip that keeps it secured in lab coat pockets.
UOM: 1 * 3 items
Supplier: HUPFER METALLWERKE
Description: Stainless steel or steel, coated.
Sale
Supplier: BOCHEM
Description: These handles are made of stainless steel 18/10 and are used to carry glass beakers.
Catalog Number: (139-3120)
Supplier: Thermo Fisher Scientific
Description: HDPE. Foam inserts.
UOM: 1 * 1 items
Supplier: Thermo Fisher Scientific
Description: These bottle carriers allow for safe transport and storage of equipment, such as bottles, and containers. They include a partition wall and an integra...
Supplier: Bel-Art Products, a Part of SP
Description: HDPE.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 37
no targeter for Bottom