You Searched For: Cables
44  results were found
SearchResultCount:"44"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Avantor Fluid Handling
Description: Create your own cable for optimization of your Masterflex® drive.
Supplier: Avantor Fluid Handling
Description: Easily connect your Masterflex® drive to a PLC or other control device.
Supplier: Avantor Fluid Handling
Description: Increase modular drive flexibility with extended cable lengths.
Supplier: VWR Collection
Description: Cable, Lemo (metrohm ) connector, 1 m cable, coaxial, Coax57, Length: 1 m
Supplier: Avantor Fluid Handling
Description: Displays instantaneous, total and accumulated flow rates.
Supplier: Avantor Fluid Handling
Description: Optimize performance of your Masterflex® MasterSense™ or Ismatec® Reglo Touchscreen pump.
Catalog Number: (444-2853)
Supplier: VWR Collection
Description: Cable, IEC power cord, Euro plug
UOM: 1 * 1 items
Catalog Number: (MFLX22050-54)
Supplier: Avantor Fluid Handling
Description: Optimize performance of your Masterflex® pump.
UOM: 1 * 1 items
Catalog Number: (MFLX77095-02)
Supplier: Avantor Fluid Handling
Description: Make the most of your Liqui-Sense leak detector with suitable replacement parts.
UOM: 1 * 106,68 cm
Catalog Number: (MFLX22050-58)
Supplier: Avantor Fluid Handling
Description: Optimize performance of your Masterflex® pump.
UOM: 1 * 1 items
Catalog Number: (700-0853)
Supplier: VWR Collection
Description: Replacement cables suitable for all PerfectBlueâ„¢ electrophoresis systems and blotting systems, and are compatible with the majority of commercially av...
UOM: 1 * 1 PAIR
Catalog Number: (MFLX22050-60)
Supplier: Avantor Fluid Handling
Description: Optimize performance of your Masterflex® pump.
UOM: 1 * 304,8 cm
Supplier: WTW
Description: Cable, AS/IDS-25, connection cable for IDS sensors with waterproof plug head, Length: 25 m
Supplier: Bohlender
Description: PTFE, 4-pole measuring cable and Lemo coupling. It is suitable for all BOLA temperature probes with coupling type Lemo, socket size 1.
Supplier: Bohlender
Description: Cables connect waste caps with electronic fill level control or sensors to a b.safe display to monitor the fill level.
Supplier: Brady
Description: Cable, Nylon cable ties, width 3,6 mm, Length: 14 cm
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 44
no targeter for Bottom