You Searched For: Buckets
59  results were found
SearchResultCount:"59"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (331-0114)
Supplier: VWR Collection
Description: Buckets made of white food grade PP.
UOM: 1 * 30 L
Supplier: BÃœRKLE
Description: HDPE, white or black.
Supplier: BÃœRKLE
Description: Made of PP, translucent.
Supplier: PFENNIG REINIGUNGSTECHNIK
Description: Standard for Clino® system trolleys, rectangular bucket with handle. Also suitable for the pre-preparation of textiles.
Supplier: BOCHEM
Description: Buckets, stainless steel 18/10.
Supplier: BÃœRKLE
Description: Made of PE, transparent.
Catalog Number: (CONT2743S)
Supplier: CONTEC
Description: Twin Bucket is an autoclavable 20 L divided bucket made of durable PP with a curved stainless steel wringer.
UOM: 1 * 1 items
Supplier: BOCHEM
Description: Stainless steel 18/10.
Supplier: BÃœRKLE
Description: Made of PP, white.
Supplier: PFENNIG REINIGUNGSTECHNIK
Description: Universal multipurpose rectangular PP buckets with handle.
Supplier: PFENNIG REINIGUNGSTECHNIK
Description: Standard bucket, rectangular, made of PP, with stainless steel handle, for Clino® system trolleys.
Supplier: SciLabware
Description: HDPE, white or black.
Supplier: RSG
Description: Stainless steel, rust-proof, polished finish.
Supplier: Brand
Description: PP, translucent.
Supplier: Kautex
Description: HDPE, with graduation.
Supplier: USBECK Laborgeräte
Description: Stainless steel.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 59
no targeter for Bottom