VWR®, Cleaning Brushes for Bottles
Supplier: VWR
VWR®
149-0207EA 29.4 EUR
149-0207 149-0205 149-0206
VWR®, Cleaning Brushes for Bottles
Brushes
Black bristles with wire handle. The special design enables bottles to be cleaned thoroughly and effectively.
  • Bristles flatten together and spread out again when they have passed through the bottle neck
  • Double-tied tip minimises risk of scratching and improves cleaning intensity
  • Hanging loop