VWR®, Cleaning Brushes for Small Vessels
Supplier: VWR
VWR®
149-0222EA 22.9 EUR
149-0222 149-0216 149-0217
VWR®, Cleaning Brushes for Small Vessels
Brushes
Brushes with white nylon or natural bristles and wire handle. Ideal for cleaning reagent flasks, burettes, pipettes, tubes and a wide variety of other laboratory items with small diameters.
  • Available with white nylon or natural bristles
  • Wire handle for secure grip
  • Hanging loop