VWR®, Cleaning Brushes for Glass Beakers
Supplier: VWR
VWR®
149-0134EA 42.1 EUR
149-0134
VWR®, Cleaning Brushes for Glass Beakers
Brushes
Brushes with white nylon bristles and wooden handle. Ideal for intensive, extra-thorough cleaning of glass beakers and a wide range of other wide necked laboratory utensils.
  • Chemical resistant bristles
  • Strong tufts at the brush tip reduce the risk of scratching and improve cleaning intensity
  • Hanging loop