VWR®, Cleaning Brushes for Bottles
Supplier: VWR
VWR®
149-0138EA 38.2 EUR
149-0138
VWR®, Cleaning Brushes for Bottles
Brushes
Natural black bristles with wire handle.
  • Rounded tip for gentle, thorough cleaning
  • Handle with safety ribbing for a firm grip when cleaning glass equipment
  • Hanging loop