VWR®, Cleaning Brushes for Burettes
Supplier: VWR
VWR®
149-0215EA 20.4 EUR
149-0215
VWR®, Cleaning Brushes for Burettes
Brushes
Brushes with natural white bristles and wire handle. Ideal for cleaning burettes, pipettes and long tubing.
  • Long, flexible handle
  • Fan tip prevents scratching
  • Hanging loop