VWR®, Cleaning Brushes for Test Tubes
Supplier: VWR
VWR®
149-0218EA 18.6 EUR
149-0218 17145-005 149-0208 149-0219 149-0220 149-0213 149-0221 149-0214
VWR®, Cleaning Brushes for Test Tubes
Brushes
White or natural bristles. Ideal for cleaning test tubes and a wide range of other laboratory utensils.
  • One or two tip ties minimise risk of scratching and improve cleaning intensity
  • Wire handle for secure grip
  • Hanging loop