You Searched For: Brushes
61  results were found
SearchResultCount:"61"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Q Path NovaBrush® are disposable brushes designed for endo- and exocervical cytological sampling. The shape of Q Path® NovaBrush 1's head makes it ide...
Sale
Supplier: DESCO
Description: Brushes minimise static charge generation and remove electrostatic charges to ground when held by grounded personnel.
Supplier: PFENNIG REINIGUNGSTECHNIK
Description: Joint scrubber/brush for gaps in hygienic sensitive areas.
Catalog Number: (PPSA3590500)
Supplier: PFENNIG REINIGUNGSTECHNIK
Description: Handy brush with long handle for hygienic sensitive areas.
UOM: null
Supplier: Sampling Systems
Description: Brushes with nylon bristles in a twisted stainless steel wire frame and aluminium handle. Ideal for cleaning both stainless steel and plastic samplers...
Supplier: MICRONOVA
Description: The FlexBrushâ„¢ is ideal for cleaning over and around piping and fittings. The flexibility of the head allows the user to shape the head for custom cle...
Waste Reducing
Supplier: DELTALAB
Description: These cytology brushes are ideal for cervical specimen collection, designed for taking cell samples without damaging them.
Catalog Number: (MICRBRWH-50)
Supplier: MICRONOVA
Description: Round head brush features medium stiff polyester bristles for aggressive cleaning.
UOM: 1 * 1 items
Supplier: MICRONOVA
Description: Brushes with polyester bristles and plastic handle for cleaning bottles and other glassware.
Supplier: MICRONOVA
Description: These durable, 100% polypropylene brushes clean finely polished stainless steel parts without scratching.
Supplier: GERBER FUNKE DR N.
Description: Brush, Cleaning brush for butyrometer body, For: Butyrometer, Handle material: Wire, Total length: 270 mm
Catalog Number: (MICRBRWM-345)
Supplier: MICRONOVA
Description: Stainless steel wire brushes with nylon bristles.
UOM: 1 * 12 items
Catalog Number: (SCOT290740059)
Supplier: DWK Life Sciences
Description: Stubborn soiling and deposits in NMR tubes can be easily removed with the NMR brush.
UOM: 1 * 1 items
Supplier: MICRONOVA
Description: The kettle brush is an oblong-shaped brush with a flare head design and polyester bristles, that is well suited for an all-around, thorough cleaning o...
Catalog Number: (MICRBRWH-66)
Supplier: MICRONOVA
Description: This wirewound brush features a smaller diameter handle and softer bristles for delicate cleaning of jars and bottles with smaller openings.
UOM: 1 * 1 items
Supplier: MICRONOVA
Description: A stiff-bristle brush head and threaded plastic handle, specifically designed for cleaning floor drains. The brush head features black bristles for co...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 61
no targeter for Bottom