You Searched For: Waste Disposal Boxes
13  results were found
SearchResultCount:"13"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (212-1833)
Supplier: VWR Collection
Description: This acrylic box is ideal for the one-handed disposal of used pipette tips. The 10 mm thick acrylic provides protection from β radiation.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Corrugated cardboard with PE pouch, 50 µm thick.
Supplier: ECONIX LTD
Description: These U.N. approved, paper-based containers provide a safe and innovative way to dispose all kinds of waste.
Supplier: ECONIX LTD
Description: The 6 L Bio-bin® has been specifically designed to safely contain serological pipettes in labs.
Catalog Number: (HUAROND-ANDEC12N)
Supplier: Hospidex
Description: Double-sided corrugated cardboard box.
UOM: 1 * 10 items
Supplier: CEMO GMBH
Description: These mobile boxes with its stable design enables clean and safe storage of hazardous solid goods belonging to packing groups II or III and is also ap...
Supplier: HAMILTON BONADUZ
Description: The box is used with waste chute 736-0847 and 736-0846.
Catalog Number: (MARO108000.00)
Supplier: MARTOR ARGENTAX
Description: This box is designed for the safe deposition of 'used' snap-off blade segments, blade ends, straight/hook/industrial cutter/deburring scalpel and hobb...
UOM: 1 * 1 items
Catalog Number: (129-4288)
Supplier: DELTALAB
Description: PP, yellow.
UOM: 1 * 1 items
Supplier: Bel-Art Products, a Part of SP
Description: Corrugated cardboard.
Supplier: Allpack srl
Description: High quality corrugated cardboard with polyethylene bag.
Catalog Number: (129-1300)
Supplier: Allpack srl
Description: Cardboard box, PE bag.
UOM: 1 * 10 items
Supplier: Roth Carl
Description: Disposal boxes are made of cardboard, with 50 μm thick polyethylene liner bag, which prevents leakage of liquid residues.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 13 of 13
no targeter for Bottom