You Searched For: Storage Boxes
99  results were found
SearchResultCount:"99"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: This range of plastic boxes offers a modern and practical solution for the storage, organization, transportation, and presentation of small parts. Wit...
New Product Sale
Supplier: VWR Collection
Description: This range of euro containers are made entirely from recycled polypropylene and adhere to the guidelines of the 'Der Blaue Engel' ecological label. Su...
New Product Sale
Supplier: VWR Collection
Description: PS, clear.
Catalog Number: (631-1067)
Supplier: VWR Collection
Description: Filing cabinet in kraft cardboard.
UOM: 1 * 10 items
Catalog Number: (631-1072)
Supplier: VWR Collection
Description: Storage organiser with two metal drawers to store embedding cassettes.
UOM: 1 * 1 items
Catalog Number: (631-1050)
Supplier: VWR Collection
Description: Storage organiser with 2×4 metal drawers for embedding cassettes.
UOM: 1 * 1 items
Catalog Number: (631-1065)
Supplier: VWR Collection
Description: Filing cabinet in white cardboard.
UOM: 1 * 2 items
Supplier: MULTIROIR
Description: This range consists of four storage bins, all made from 100% recycled polypropylene.
New Product Sale
Supplier: MULTIROIR
Description: PP, translucent.
Catalog Number: (BRDY065241)
Supplier: Brady
Description: These enclosed modules provide safe storage and control access to padlocks.
UOM: 1 * 1 items
Supplier: MULTIROIR
Description: Foldable containers made of black PP with 'active lock' system for fast, easy and safe operation. These folding containers are light weight and very ...
Supplier: MULTIROIR
Description: These storages boxes are a premium organising system developed with a vision of functionality, high quality, and a clear Scandinavian design.
Supplier: MULTIROIR
Description: The multiroir dividable plastic storage box allows the storage or organisation of products on shelves or on other supports.
Supplier: MULTIROIR
Description: These airtight boxes are designed to withstand the harshest conditions, both indoors and outdoors. When tightly closed, the boxes become completely se...
New Product
Supplier: MULTIROIR
Description: Made of PS.
Supplier: MULTIROIR
Description: The MLT series plastic storage bins are designed to optimize storage, organization, and picking in all sectors of activity.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 99
no targeter for Bottom