You Searched For: Cryoboxes
95  results were found
SearchResultCount:"95"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Cardboard, coated.
Supplier: VWR Collection
Description: Corrugated cardboard box.
Supplier: VWR Collection
Description: Made of extra strong polycarbonate.
Supplier: VWR Collection
Description: Cardboard, coated.
Supplier: VWR Collection
Description: Cardboard, white.
Supplier: VWR Collection
Description: Cardboard.
Supplier: VWR Collection
Description: PP.
Supplier: VWR Collection
Description: Cryo boxes are suitable to store up to –196 °C.
Supplier: VWR Collection
Description: Boxes, made of cardboard, white. For storage of cryovials at low temperatures.
Catalog Number: (479-2214)
Supplier: VWR Collection
Description: Cryoboxes, fiberboard, sturdy, water-repellent, with covers for sample storage.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Made of white cardboard.
Supplier: VWR Collection
Description: Cardboard, coated.
Supplier: VWR Collection
Description: Corrugated cardboard boxes.
Supplier: VWR Collection
Description: Made of corrugated or thin film PP.
Catalog Number: (478-0241)
Supplier: VWR Collection
Description: Polycarbonate.
UOM: 1 * 1 items
Catalog Number: (479-1349)
Supplier: VWR Collection
Description: Made from corrugated PP.
UOM: 1 * 10 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 95
no targeter for Bottom