Bottles, wide neck, square, with screw cap
Supplier: Gosselin
215-3254EA 168 EUR
215-3254 212-5185 215-3956 GOSSLC2000-08 215-3952 GOSSF2700-06 215-1292 215-3248 215-0722 215-3951 GOSSLC2000-04 215-3251 215-0719 215-3954 GOSSLC2500A-01 215-0718 215-3255 GOSSLC2000-17 GOSSLC2500A-04 215-3271 215-0710 215-1291 GOSSLC2500A-08 GOSSLC2500A-02 215-3929 GOSSLC2500A-09 215-3950 GOSSLC1000-03 215-0708 215-0720 GOSSLC2000-07
Bottles, wide neck, square, with screw cap
Bottles Plastic Bottles
Space saving bottles made of HDPE, translucent with HDPE or LDPE screw cap.
  • Good chemical resistance, good temperature resistance
  • Available sterile or packed under aseptic conditions
  • Excellent shock resistance
Ideal for liquid, solid and powder storage. Good level of tightness according to the closure system used and application.