You Searched For: Cell Culture Bottles
35  results were found
SearchResultCount:"35"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: PC, autoclavable, narrow mouth bottles with standard PP caps.
Supplier: VWR Collection
Description: Square PETG bottles are ideal for storage of media, laboratory reagents and biological buffers up to −70 °C. Eliminate washing, depyrogenation and aut...
New Product Sale
Supplier: VWR Collection
Description: Sterile polycarbonate containers, with weldable tubing, provide the maximum amount of flexibility and reliability. These vessels are assembled in an I...
Sale
Supplier: VWR Collection
Description: PS, PC or PETG, with PP screw cap, sterile.
Sale
Supplier: VWR Collection
Description: Sterile PETG containers, with weldable tubings, provide the maximum amount of flexibility and reliability. These vessels are assembled in an ISO Class...
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
Supplier: Thermo Fisher Scientific
Description: Certified clean container is made of PETG and comes with a sterile, natural HDPE closure with a tamper-resistant seal.
Supplier: Thermo Fisher Scientific
Description: PETG.
Sale
Supplier: Thermo Fisher Scientific
Description: PETG, clear with silicone/PTFE screw cap.
Supplier: Thermo Fisher Scientific
Description: PETG bottle with HDPE screw closure.
Sale
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
Supplier: Thermo Fisher Scientific
Description: Square shaped media bottles.
Supplier: Thermo Fisher Scientific
Description: PET, clear, with thread or complete with white HDPE screw caps.
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear, with porcelain clamp closure and rubber seal.
Catalog Number: (391-0690)
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
UOM: 1 * 1 items
Supplier: Thermo Fisher Scientific
Description: PETG bottle with HDPE screw closure.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 35
no targeter for Bottom