You Searched For: Blotting Papers
15  results were found
SearchResultCount:"15"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: This super thick paper is used to complete the blotting sandwich. Smooth, uniform texture ensures continuous contact of buffer from the chamber throug...
Supplier: VWR Collection
Description: Ideal for use as wicks in protein and nucleic acid blotting. Provides a uniform flow of buffer through the gel to the transfer membrane in a blotting ...
Supplier: OWL SCIENTIFIC
Description: Blotting paper, Owl™, for semi-dry electroblotting, Cotton, Sheets, 100×100 mm, Weight: 183 g/m², Thickness: 0,83 mm
Supplier: Sartorius
Description: Their extremely smooth surface and relatively high thickness provide them with superior absorption properties and uniformity across the entire contact...
Supplier: Thermo Fisher Scientific
Description: These Western blotting filter papers are pre-cut cotton sheets for wet or semi-dry, passive or electrophoretic transfer of proteins from polyacrylamid...
Supplier: Ahlstrom-Munksjö
Description: Ahlstrom-Munksjö offers a full range of high quality cotton papers with high absorbency – ideal for chromatographic analysis and blotting methods with...
Supplier: Thermo Fisher Scientific
Description: Filter paper designed specifically for Western blotting.
Supplier: Whatman products (Cytiva)
Description: This general purpose blotting paper is made from pure raw materials with a high absorbency and is used as a membrane gel support. Due to its thickness...
Catalog Number: (515-8110)
Supplier: Whatman products (Cytiva)
Description: Absorbent paper strips made from cellulose for the removal of excess liquids, e.g., from microscope specimens.
UOM: 1 * 100 items
Supplier: Whatman products (Cytiva)
Description: Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
Catalog Number: (631-0866)
Supplier: GLASWARENFABRIK KARL HECHT
Description: Blotting paper, Spare for no. 1238, Sheets, 200×45 mm
UOM: 1 * 25 items
Catalog Number: (RPN6101M)
Supplier: Cytiva
Description: Research laboratory blotting paper for use in gel electrophoresis in all molecular biology and protein studies applications.
UOM: 1 * 100 items
Supplier: Cytiva
Description: Blotting paper, Sheets, 210×260 mm
Supplier: Cytiva
Description: Blotting paper, Sheets, 145×215 mm
Supplier: Roth Carl
Description: Blotting paper, Whatman 3MM, Cellulose, Sheets, 200×200 mm, Thickness: 0,34 mm
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 15 of 15
no targeter for Bottom