You Searched For: Adapters
125  results were found
SearchResultCount:"125"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Ring adapter for mono- and binocular microscopes (Ø = 30 mm)
Supplier: VWR Collection
Description: PTFE, with ground joints on cone and socket.
Supplier: VWR Collection
Description: C-mount camera adapter for Olympus microscopes
Supplier: VWR Collection
Description: Borosilicate 3.3 glass.
Supplier: VWR Collection
Description: Eyepiece adapter, 0,45x, Ø: 23/30 mm
Supplier: VWR Collection
Description: These adapters enable cameras with different chip sizes to be connected to microscopes with a C-mount thread.
Catalog Number: (630-1747)
Supplier: VWR Collection
Description: Phototube adapter for digital camera
UOM: 1 * 1 items
Supplier: VWR Collection
Description: PTFE, strong walls.
Supplier: VWR Collection
Description: C-mount adapter compatible with Nikon microscopes, 0,5×
Catalog Number: (MFLX06432-00)
Supplier: Avantor Fluid Handling
Description: Forms a continuous standard taper inside and out, from top to bottom.
UOM: 1 * 1 items
Supplier: DWK Life Sciences
Description: Made of DURAN®, borosilicate glass 3.3, clear.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Made of DURAN®, borosilicate glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Made from DURAN® tubing.
Supplier: Bohlender
Description: For assembly of components with ground joint cone NS 29/32 on bottles and vessels with GL 45 thread.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 125
no targeter for Bottom