You Searched For: Rectangular Tanks
9  results were found
SearchResultCount:"9"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: BÃœRKLE
Description: These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.
Catalog Number: (ENTEE92-01D-0215)
Supplier: ENTEGRIS INC
Description: Injection molded rectangular tanks with dual capacity.
UOM: 1 * 1 items
Supplier: Thermo Fisher Scientific
Description: These PP rectangular tanks with cover can significantly reduce evaporation and contamination.
Supplier: Thermo Fisher Scientific
Description: These heavy-duty rectangular LLDPE tanks with cover can withstand most acid, alcohols and bases. Tanks feature stepped flange at top provides reinforc...
Catalog Number: (CEMG7536)
Supplier: CEMO GMBH
Description: Tank from HDPE with general construction inspection approval with rigidly connected sheet steel pallet for handling with a forklift or pallet truck fr...
UOM: 1 * 1 items
New Product
Supplier: Bel-Art Products, a Part of SP
Description: These seamless, rotational-moulded PE tanks are lightweight and easy-to-handle.
Supplier: GLASWARENFABRIK KARL HECHT
Description: Tubs made of Plexiglas®, available with or without sloping base. Sloping base facilitates circulation of the liquid.
New Product
Supplier: SEMADENI
Description: These tanks can be used for a long time at temperatures of +50 °C.
Catalog Number: (SMIT1195/LID)
Supplier: SMITH SCIENTIFIC LTD
Description: Chromatography tank made up of clear, soda-lime glass.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 9 of 9
no targeter for Bottom