Portable balances, Navigator® and Navigator® XL series
Supplier: Ohaus
Navigator
611-2843EA 265 EUR
611-2843 OHAU83033085 611-5403
Portable balances, Navigator® and Navigator® XL series
Balances and Scales
Two series of lightweight, robust, portable balances that are ideal for use in a wide range of laboratory and industrial applications. The innovative, touch-free sensors allow up to 36 combinations of functions including tare, zero, print, function and, where appropriate, display backlight. The applications include weighing (with 14 units), percentage weighing, check weighing, parts counting, accumulation/totalisation and display hold. The calibration function is digital and requires an external weight.
  • Dual multi-functional touch-free sensors free up your hands, keeping the balance free of contaminants delivering speed, safety and cleanliness
  • A polished stainless steel pan, rugged, corrosion-free plastic housing and spill-resistant pan sub-assembly embodies a hygienic design that is easy to clean, IP 43
  • Fast; stabilises in less than one second
  • A bright, backlit LCD with 20 mm digits provides optimum visibility in any lighting condition
  • The mechanical overload protection system enables the scale to safely bear loads up to four times its rated capacity
The non slip adjustable feet and the integral levelling bubble indicator, which is illuminated at power-up, help the operator ensure precision and reproducibility wherever the balance is used.
Ordering information: A full range of accessories and also EC type approved models for Navigator® range are also available – please ask Avantor for details. An optional internal rechargeable battery option is available for the Naviagator® XL range.
Delivery information: Supplied with AC adapter (NV models require 4× AA (LR6) batteries, not supplied; NVL models require 4× C type (LR14) batteries, not supplied).