Microarray printing plates
Supplier: Corning Life Sciences
734-4119EA 0 EUR
CORN3672
Microarray printing plates
PP, non sterile. Low volume microplates meeting source plate requirements for printing DNA content onto microarray slides. The plates feature rigid, full length skirts for full compatibility with automation. Full volume version also available, see Cat. No. 732-5528.
  • Conical V-bottom well design with square well geometry provides maximum sample recovery
  • Resistant to many organic solvents, including DMSO
  • Certified DNase- and RNase-free
  • Automation compatible
Recommended working volume: 2 - 10 µl