Nutrient media steriliser, MediaClave 10
Supplier: Integra Biosciences
MediaClave
710-0734EA 0 EUR
710-0734 INTG136005 710-0735
Nutrient media steriliser, MediaClave 10
Media Preparators
The MediaClave 10 culture media steriliser for 1 to 10 L of culture media features an impressive compact design and state-of-the-art technology while remaining simple to operate. Thanks to its efficient, innovative continuous-flow heater, powerful magnetic stirrer and rapid cooling process, the MediaClave 10 can handle the demands of any material, be it standard agar, viscous media or highly sensitive media. It is consistent with the latest device documentation and of course conforms to GMP/GLP and FDA CFR21 Part 11. Steriliser, bench-top autoclave and water bath functions in one instrument with automatic cleaning program.
  • Compact bench-top device
  • Highly efficient rapid cooling process
  • TÜV/CE certified
  • LCD display
  • 50 programs
  • Ethernet interface/WebServer