Centrifuge, refrigerated, floor-standing, Sorvall™ WX80 / WX100 Ultra
Supplier: Thermo Fisher Scientific
Sorvall™
521-1619EA 57750 EUR
521-1619 SORV46902
Centrifuge, refrigerated, floor-standing, Sorvall™ WX80 / WX100 Ultra
Centrifuges Floor-Standing Centrifuges
The Sorvall WX Ultracentrifuge Series delivers the speed and processing power to meet high-throughput processing needs.
  • Intuitive colour touch LCD screen, up to 20 programmes stored into memory
  • UltraSecur software enabling rotor life management and controlled access
  • Small footprint, quiet run, imbalance-tolerant drive
  • Easy sample loading for safer separations
  • Simulation software allowing development of separation protocols on laptop or PC and ensuring regulatory compliance
Typical applications: Cell lysate clarification, subcellular organelle/plasma membrane isolations, genomic DNA/Plasmid DNA/RNA isolation/purification, porotein purification, bacteria/yeast/virus pelleting.
Ordering information: Supplied without rotors.