Portable balances, CL series
Supplier: Ohaus
CL Series
611-1875EA 55 EUR
611-1875 611-4995 611-4996
Portable balances, CL series
Balances and Scales
The CL series balance is a lightweight, portable scale perfectly suited for many applications including quality control, formulation, soil sampling, jewellery, postal weighing and dietary weighing. Choice of displayed units; gram, pound, ounces, ounce troy, pennyweight.
  • Compact design and storage made easy with a stackable design
  • ABS housing and pan
  • Integrated load cell lock
  • Battery operation with auto shut-off
  • External calibration
Delivery information: Supplied with 3×AA batteries. A/C adapter is available but not supplied, please contact your local Avantor sales office for details.