Minisart® syringe filters NML
Supplier: Sartorius
516-0903EA 798 EUR
516-0903 512-3118 514-7013 514-7012 611-0708 514-7011 514-7014 611-0707 611-0691 514-0674 514-0673 514-0672
Minisart® syringe filters NML
Filters Syringe Filters
Minisart® NML with surfactant-free cellulose acetate (SFCA) membrane is the best choice for all aqueous solutions and oils with a pH of 4 to 8. The SFCA membrane is the best option for leachable-free filtrations. The very low non specific binding characteristics of this membrane make it a perfect choice for biomolecule solutions.
  • PVC-free
  • 100% optical integrity test
  • Colour-coded MBS housing for easy pore identification
  • Superior flow rate
  • Bidirectional use possible
The housing is made of the acrylic resin MBS (Methacrylate Butadiene Styrene) which has a limited compatibility with alcohols but is free of phthalates and passes the plastic class VI test. Housing and membrane are tightly connected which allows bidirectional use.
Individually packed and sterile Minisart® NML syringe filters are optimal for immediate usage whenever sterile filtration or clarification and cell removal under sterile conditions is needed.