You Searched For: Pressure-Relief Valves
13  results were found
SearchResultCount:"13"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Bohlender
Description: Body made of PTFE with two connections with female thread UNF 1/4" 28 G. Valve made of PPS with set screw and lock nut for adjusting and fixing pressu...
Supplier: ENTEGRIS INC
Description: These components are constructed of high purity PFA and PTFE materials for wetted parts and PVDF, PEEK™ and PTFE u-cup with Viton® energizer for non w...
Supplier: Avantor Fluid Handling
Description: Prevent leaks with the emergency shutdown design.
Supplier: Bohlender
Description: Pressure compensation sets consisting of screw cap made of PPS for GL thread, PTFE/silicone gasket and exchangeable filtering membrane made of PTFE. F...
Catalog Number: (MFLX01350-82)
Supplier: Avantor Fluid Handling
Description: Easily control your outlet pressure.
UOM: 1 * 1 items
Catalog Number: (MFLX03245-40)
Supplier: Avantor Fluid Handling
Description: Easily control your outlet pressure.
UOM: 1 * 1 items
Catalog Number: (201-0290)
Supplier: Bohlender
Description: Valve made from PPS with flow body made from PTFE. Can be used as a pressure relief valve with adjustable control of pressure or as a valve to prevent...
UOM: 1 * 1 items
Supplier: Welch by Gardner Denver
Description: Small flange valve, open type, made of nickel-plated aluminium.
Catalog Number: (119-0234)
Supplier: JUSTRITE MANUFACTURING CO
Description: These safety drum vents provide automatic pressure relief for drums between 0,20 bar but less than 0,55 bar. Depending on the model, vacuum relief is ...
UOM: 1 * 1 items
Catalog Number: (BROE96T0227)
Supplier: BROEN-LAB
Description: Pressure reducing valve 2,0 for Broen showers, 0,5 - 8,5 bar
UOM: 1 * 1 items
Supplier: Restek
Description: These valves and tees are made from stainless steel or are Sulfinert®-treated.
Supplier: Restek
Description: Valves for ASE® 100/200/300 systems.
Catalog Number: (VARIRE7030)
Supplier: VARIAN
Description: Valves made from stainless steel are two-position with six-port. It is manually operated with three-way switching.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 13 of 13
no targeter for Bottom