You Searched For: Nessler Tubes
8  results were found
SearchResultCount:"8"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: DWK Life Sciences
Description: The Nessler tubes, with shadowless bottom that transmits undistorted light, are ideal for colour comparison. Rings and markings are in durable white c...
Catalog Number: (SCERSP45310-50100)
Supplier: DWK Life Sciences
Description: Manufactured from 33 expansion, low extractable borosilicate glass.
UOM: 1 * 6 items
Supplier: MARIENFELD
Description: Nessler cylinder, low form, with white circular mark at 50 ml
Supplier: WITEG LABORTECHNIK GMBH
Description: Colour comparison cylinders according to Nessler for comparators, with optically plane bottom.
Supplier: WITEG LABORTECHNIK GMBH
Description: Eggertz tubes, 20 ml with 0,1 ml division for comparators
Supplier: SciLabware
Description: These Nessler cylinders are designed for colour comparison of water samples, foodstuffs, etc
Supplier: NEUBERT VOLUM. GLASSWAERE
Description: Nessler cylinders, 100 ml, tall form, Ø×H: 26×320, 50 + 100 ml division
Catalog Number: (HACH173606)
Supplier: Hach
Description: Tube with 5 and 20 ml marks for test kit DE2
UOM: 1 * 6 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8 of 8
no targeter for Bottom