You Searched For: Microplate Racks
13  results were found
SearchResultCount:"13"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: TENAK AS
Description: Complete range of racks for microtiter plates for use in upright freezers, made of stainless steel. The racks can either be used as one plate per shel...
Supplier: TENAK AS
Description: Microplate racks, made of aluminium, can be used for all freezer models.
Supplier: TENAK AS
Description: These microplate racks, made of aluminium, can be used for all freezer models.
Supplier: TENAK AS
Description: Stainless steel standard microplate racks.
Supplier: TENAK AS
Description: This is a complete range of racks for microtiterplates for use in chest freezers.
Supplier: Thermo Scientific
Description: Maximise the storage space in your ultra-low temperature freezer with these Thermo Scientificâ„¢, racks for Revcoâ„¢ ExF, DxF and HERAfeezeâ„¢ HFU B freezer...
Catalog Number: (HERA398188)
Supplier: Thermo Scientific
Description: Use Thermo Scientificâ„¢ chest freezer racks with TDE series ultra-low temperature chest freezers as well as legacy Formaâ„¢, HERAfreezeâ„¢, Revcoâ„¢ and TS-s...
UOM: 1 * 1 items
Supplier: Thermo Scientific
Description: Maximise tube storage capacity per freezer with Thermo Scientificâ„¢ Sliding Drawer Racks for Tubes (5 inner door freezers), a racking solution for five...
Catalog Number: (NALG5039-0048)
Supplier: Thermo Fisher Scientific
Description: Stainless steel.
UOM: 1 * 1 items
Supplier: GLW
Description: These racks are commonly used for chest freezers.
Supplier: Thermo Fisher Scientific
Description: A range of storage racks for standard microplates and DeepWellâ„¢ plates in chest or upright mechanical freezers or incubators.
Supplier: GLW
Description: Stainless steel
Catalog Number: (REVC398188)
Supplier: REVCO TECHNOLOGIES
Description: Rack, Microplate storage/freezer, For: Microplates, Shelves/drawers per rack: 27, 135×89×655 mm
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 13 of 13
no targeter for Bottom