Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "preprinted-labels"
https://ru.vwr.com/stibo/smallweb/std.lang.all/53/39/18945339.jpg
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"5"
  List View Grid View Easy View (new)
Sort by:
Supplier:  BRADY & CO LTD, W. H.
Description:   These two part tags provide enhanced operational control and record keeping. Matching identification numbers are printed on each part.
Supplier:  BRADY & CO LTD, W. H.
Description:   These two-part tags provide enhanced operational control and record keeping.
Supplier:  EP SCIENTIFIC
Description:   Label is colour-coded with the chemical preservative type for quick reference and safe handling. Sticks securely to plastic or glass sample containers...
Catalog Number: (DAKL464085)
Supplier:  DAKLAPACK
Description:   LABEL DRY ICE UN1845 WHITE 1 * 500 items
UOM:  1 * 500 items
Catalog Number: (DAKL460610)
Supplier:  DAKLAPACK
Description:   This pre printed labels are 100% compliant with the UN3373 packaging laws.
UOM:  1 * 100 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5  of 5