You Searched For: Gowning Racks
13  results were found
SearchResultCount:"13"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Palbam
Description: Gown racks in stainless steel 304L with electropolished finish
Supplier: Palbam
Description: These shorter gown racks are designed for hanging lab coats and frocks.
Supplier: Metro International
Description: Racks are designed to provide efficient storage of cleanroom garments. The hangers are spaced at 75 mm intervals.
Supplier: Metro International
Description: Hanger tubes in stainless steel attach easily under Super Erecta shelves for hanging garments.
Supplier: Palbam
Description: Wall-mounted gownracks in stainless steel 304 with electropolished finish.
Catalog Number: (PALBGC1302)
Supplier: Palbam
Description: Cleanroom cubby racks.
UOM: 1 * 1 items
Catalog Number: (HUPF7500452)
Supplier: HUPFER METALLWERKE
Description: This shoe rack is made of stainless steel with electropolished finish. It can hold 6 pairs of shoes or clogs suspended and also it can be fitted to wa...
UOM: 1 * 1 items
Supplier: TEKNOMEK
Description: Our hygienically designed stainless steel shoe and boot racks come with a hygienic, easier to clean, continuous piece of prongs.
Supplier: TEKNOMEK
Description: Our stainless-steel hanging racks are manufactured to the highest standards.
Supplier: TEKNOMEK
Description: Our hanging racks are designed for standard portrait clipboards.
Supplier: TEKNOMEK
Description: Open hook garment hangers are manufactured from 304 grade stainless steel in a polished finish.
Supplier: TEKNOMEK
Description: Our free-standing rail can be used either with coat hangers or with hooks.
Catalog Number: (SCAA460014)
Supplier: Essity
Description: The Tork coat hook is made of stainless steel with anti-finger print coating that looks great and stays clean.
UOM: 1 * 10 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 13 of 13
no targeter for Bottom