Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Evacuation Equipment
https://ru.vwr.com/stibo/smallweb/std.lang.all/95/64/18709564.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"3"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (111-1375)
Supplier:  HONEYWELL SAFETY
Description:   This triangle is suitable for short-duration rescue and evacuation operations of an uninjured casualty in a sitting position.
UOM:  1 * 1 items
Catalog Number: (DALL1002920)
Supplier:  HONEYWELL SAFETY
Description:   Pivot davit column, portable, uni-hoist system with integrated 30 m rescue winch, H-base and adjustable legs, W×D×H: 1300×1300×2250 (fully extended) mm
UOM:  1 * 1 items
Catalog Number: (111-1387)
Supplier:  HONEYWELL SAFETY
Description:   Lightweight body splint that is ideal for confined space evacuation. Once wrapped around a casualty, it provides a rigid splint. Ideal for the army an...
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 3  of 3