You Searched For: First Aid Kit Components
10  results were found
SearchResultCount:"10"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (PLUM5151)
Supplier: Plum Safety
Description: These wound cleansing wipes contain sterile water. Ideal for small wounds and cuts.
UOM: 1 * 20 items
Catalog Number: (PLUM4554)
Supplier: Plum Safety
Description: This spray provides immediate and effective care to minor accidents such as cuts and scrapes on the skin or dust and dirt in the eyes. It helps to min...
UOM: 1 * 1 items
Catalog Number: (118-0057)
Supplier: ORKLA CARE AB
Description: Refill pack for the first aid panel, for use in first aid situations.
UOM: 1 * 1 items
Supplier: ORKLA CARE AB
Description: Wound cleansers do away with cotton wool and bottles of antiseptic. These sterile, non woven tissues, which are saturated in a wound cleansing fluid (...
Catalog Number: (MMMEM82002A)
Supplier: 3M MEDICAL
Description: This casting tape consists of a knitted fiberglass fabric impregnated with polyurethane resin. Exposure to moisture or water initiates a chemical reac...
UOM: 1 * 10 Roll
Catalog Number: (118-0247)
Supplier: ORKLA CARE AB
Description: This cleansing spray is intended for rinsing and cleaning out dust and dirt from eyes and wounds. The portable spray bottle is filled with 150 ml phys...
UOM: 1 * 1 items
Catalog Number: (148-4191)
Supplier: Hartmann Bode
Description: Colourless antiseptic spray with ethanol, for use prior to injections, punctures and surgical procedures.
UOM: 1 * 250 mL
Catalog Number: (SAFIQ2290)
Supplier: SAFETY FIRST AID GROUP LTD
Description: This instant single use cold pack is ideal for use in the event of an emergency or injury, delivering fast first aid to those who need it. It also hel...
UOM: 1 * 1 items
Supplier: HARTMANN PAUL AG
Description: Pur-Zellin® are ready to use pre-cut pulp swabs made of highly bleached bandaging cellulose. Ideal as an absorbent pad for minor injuries, for skin cl...
Supplier: COOLIKE REGNERY GMBH
Description: The coolPack mini is a handy gel compress used for inflammations, nose bleeds, swellings, toothache and insect bites.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 10 of 10
no targeter for Bottom