You Searched For: In-Line Filters
29  results were found
SearchResultCount:"29"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (HICHHI-683)
Supplier: Avantor
Description: This is a fingertight bioanalytical inline filter, comprising a PEEK holder and a 5 μm titanium PEEK-encased frit.
UOM: 1 * 1 items
Supplier: ENTEGRIS INC
Description: These are compact, all-metal filters that fit in place of standard ¹/₄", ³/₈" and ¹/₂" VCR® gaskets in gas distribution systems. They protect sensitiv...
Supplier: SOCOREX ISBA
Description: In-line filter, Pk: 50
Supplier: Whatman products (Cytiva)
Description: Whatman Polydisc TF In-Line Filters from Cytiva are lightweight devices with high chemical and solvent resistance that are particulraly suitable for p...
Supplier: Whatman products (Cytiva)
Description: Whatman VACU-GUARD capsule filters are ideal for protecting vacuum pump systems from solvent vapor or gaseous contaminants and aqueous aerosols.
Supplier: Whatman products (Cytiva)
Description: Whatman VACU-GUARD disk filters are ideal for protecting vacuum pump systems from aqueous aerosols and water intrusion.
Supplier: Whatman products (Cytiva)
Description: Cytiva's Solvent IFD is a high-flow HPLC in-line filter/degasser, designed to simultaneously filter and degas organically based HPLC mobile phases.
Supplier: Whatman products (Cytiva)
Description: Aqueous IFD from Cytiva is a polypropylene housed in-line filter/degasser (IFD) with nylon membrane. It connects to an HPLC line and simultaneously fi...
Supplier: Whatman products (Cytiva)
Description: Polydisc HD (Heavy Duty) from Cytiva is a durable all-polypropylene in-line filter for large volumes of liquid samples containing even harsh chemical ...
Catalog Number: (516-1107)
Supplier: Whatman products (Cytiva)
Description: These disc filters contain a glass microfiber (GMF) and polyethersulfone (PES) stack designed to purify and sterilise serum and other hard to filter s...
UOM: 1 * 10 items
Supplier: Whatman products (Cytiva)
Description: Cytiva's Polydisc GW (Ground Water) is an in-line filter specifically designed for preparation of ground water samples for dissolved heavy metal analy...
Supplier: Whatman products (Cytiva)
Description: These in-line filters with PTFE membrane and PP housing are lightweight air venting units suitable for protective vents, in-line filtration and isolat...
Supplier: Whatman products (Cytiva)
Description: Whatman Polydisc AS (Aqueous Solution) in-line filters from Cytiva feature high-throughput PES membrane with a glass microfiber prefilter. Polydisc co...
Catalog Number: (SPECCU2M20512N)
Supplier: Spectrum Laboratories
Description: CultureGard® HF perfusion hollow fibre sterilising grade filter for cell culture perfusion. Made with 0,2 µm MWCO DynaFibre® membrane that permits the...
UOM: 1 * 12 items
Supplier: DWK Life Sciences
Description: These in-line filters are used for in-line filtration of gas lines to remove solid impurities (e.g. dust).
Supplier: Agilent
Description: Column inlet frit contamination that can increase column backpressure and reduce efficiency. Two types of high pressure in-line filter kits for use wi...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 29
no targeter for Bottom