You Searched For: Wire
11  results were found
SearchResultCount:"11"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: BOCHEM
Description: Steel wire gauzes with ceramic center.
Supplier: BOCHEM
Description: Triangles, steel zincked, with clay tubes.
Supplier: RSG
Description: Wire triangles made from stainless steel, with clay pipes.
Supplier: TECHNICAL QUARTZ SOLUTIONS GMB
Description: Chrome nickel wire with Vitreosil tube.
Supplier: USBECK Laborgeräte
Description: Triangles made from galvanised steel wire, with clay tubes.
Supplier: GLASWARENFABRIK KARL HECHT
Description: These wire triangles with plain clay tubes are made of galvanised steel wire.
Supplier: JUCHHEIM Laborgerate GmbH
Description: Triangles with pipeclay tubes.
Supplier: CHEMPUR
Description: Platinum 99,9%.
Catalog Number: (PERCN5320131)
Supplier: PerkinElmer
Description: Nichrome hangdown wire kit (5 hangers)
UOM: 1 * 5 items
Supplier: MURRAY, S & CO LTD
Description: Pipeclay stems on stout wire.
Supplier: CHEMPUR
Description: Platinum/Iridium 90/10 wire.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 11 of 11
no targeter for Bottom