You Searched For: Tissue Embedders
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: HERAEUS KULZER GMBH
Description: Technovit 7100 is a plastic embedding system based on HEMA (2-hydroxyethyl methacrylate). The hydrophillic resin is used in medicine, botany, zoology ...
Catalog Number: (HKUL64708805)
Supplier: HERAEUS KULZER GMBH
Description: Technovit 3040 is a quickly hardening two-component plastic with a methyl meth acrylate base (MMA).
UOM: 1 * 1 items
Catalog Number: (HKUL64715444)
Supplier: HERAEUS KULZER GMBH
Description: Technovit 9100 is a plastic embedding system used in medicine, botany and zoology.
UOM: 1 * 1 items
Supplier: Slee
Description: The MPS/P2 features ease of operation with ergonomic hand rest, spacious work surfaces, large container capacity and reliability. The extendable modul...
Supplier: Slee
Description: The MPS/P1 is a complete paraffin dispensing unit including a 3,8 litre paraffin container, a heated working area and a cold spot. The ergonomically e...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5 of 5
no targeter for Bottom