You Searched For: Chromatography Sorbents
91  results were found
SearchResultCount:"91"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Avantor
Description: The J.T.Baker® BAKERBOND® Poly Hi-Propyl mixed mode hydrophobic interaction chromatography (HIC) resin provides improved selectivity for superior sepa...
Supplier: Avantor
Description: J.T.Baker® BAKERBOND®, Diol silica irregular 40 µm, 60 Å Prep LC sorbent packing media, 100 g
Catalog Number: (7248-00)
Supplier: Avantor
Description: Prep-scale packing is intended for use at low pressure with a peristaltic pump.
UOM: 1 * 100 g
Catalog Number: (7199-00)
Supplier: Avantor
Description: J.T.Baker® BAKERBOND®, Ethyl (C₂), BAKERBOND®, 40 µm, 60 Å
UOM: 1 * 100 g
Catalog Number: (7579-01)
Supplier: Avantor
Description: J.T.Baker® BAKERBOND®, Octadecyl (C₁₈), BAKERBOND®
UOM: 1 * 1 kg
Supplier: Avantor
Description: The J.T.Baker® BAKERBOND® PolyCSX multimode ion exchange (IEX) chromatography resin functions as a strong cation exchange resin with improved selectiv...
Supplier: Avantor
Description: Silica-based column packings with high performance purification of strongly hydrophilic peptides and proteins, based on hydrophobic characteristics an...
Supplier: Avantor
Description: The J.T.Baker® BAKERBOND® PolyABx multimode ion exchange (IEX) chromatography resin functions as a weak cation exchange resin with improved selectivit...
Supplier: Avantor
Description: These are used in low pressure preparative chromatography. The mesh specifications ensure there is no excess of fines in the product which can affect ...
Catalog Number: (7061-00)
Supplier: Avantor
Description: SPE sorbent for clean-up columns. Suitable for the determination of Hydrocarbon Oil Index according ISO 9377-2 and NEN 5733.
UOM: 1 * 100 g
Supplier: Avantor
Description: Prep-scale packing is intended for use at low pressure with a peristaltic pump.
Supplier: Avantor
Description: Prep-scale packing is intended for use at low pressure with a peristaltic pump.
Supplier: Avantor
Description: The J.T.Baker® BAKERBOND® PolyPEI multimode ion exchange (IEX) chromatography resin functions as a weak anion exchange resin with improved selectivity...
Supplier: Avantor
Description: The J.T.Baker® BAKERBOND® PolyQUAT multimode ion exchange (IEX) chromatography resin functions as a strong anion exchange resin with improved selectiv...
Supplier: Avantor
Description: Packing materials for low pressure preparative and flash chromatography. Chemically stable from pH 2 to 8,5. They are not damaged by the use of cleani...
Supplier: Avantor
Description: J.T.Baker® BAKERBOND®, Silica-based reverse phase media, 15 µm, 300 Å
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 91
no targeter for Bottom