You Searched For: Buffers
630  results were found
SearchResultCount:"630"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (3861-20ML)
Supplier: SIGMA-ALDRICH MICROSCOPY
Description: Citrate buffer solution, 0.38 mol/L, pH 5.4 when diluted according to procedure 386A.
UOM: 1 * 20 mL
Catalog Number: (2851-100ML)
Supplier: SIGMA-ALDRICH MICROSCOPY
Description: These buffers are used to elute adult hemoglobin by the procedure of Oski and Naiman, a modification of the Kleihauer Betke procedure for the demonstr...
UOM: 1 * 100 mL
Supplier: SIGMA-ALDRICH MICROSCOPY
Description: For use in the Naphthol AS-D Chloroacetate Esterase Kit No. 91-C.
Catalog Number: (T6455-100ML)
Supplier: SIGMA-ALDRICH MICROSCOPY
Description: Intended for heat-induced antigen retrieval on formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies in immunohis...
UOM: 1 * 100 mL
Catalog Number: (C9999-100ML)
Supplier: SIGMA-ALDRICH MICROSCOPY
Description: Citrate buffer, antigen retriever has been used as a heat-induced antigen retriever on formalin-fixed paraffin-embedded (FFPE) tissue sections prior t...
UOM: 1 * 100 mL
Supplier: VWR Chemicals
Description: For calibration of pH meters.
Supplier: VWR Chemicals
Description: Solutions prepared from AnalaR® NORMAPUR® grade analytical reagents.
Catalog Number: (160868L)
Supplier: VWR Chemicals
Description: pH Buffer solution, Acetate, pH value: 4,50, Reference temperature: 20 °C, VWR®, Plastic container
UOM: 1 * 25 L
Supplier: VWR Chemicals
Description: These tablets provide a rapid and convenient method of preparing a buffer solution.
Supplier: VWR Chemicals
Description: Tris (hydroxymethyl) aminomethane.
Supplier: VWR Chemicals
Description: Buffer solutions, secondary standards, pH value: 12,45, ±0,05, Reference temperature: 25 °C, VWR®, Plastic bottle
Catalog Number: (E691-500ML)
Supplier: VWR Chemicals
Description: TRIS buffer solution pH 7,5 (1 mol/l), VWR®
UOM: 1 * 500 mL
Supplier: VWR Chemicals
Description: HEPES buffer solution pH 7.3 (1 mol/l), VWR®
Supplier: Avantor
Description: DILUT-IT is a ready-to-use dissolution media concentrate, which is pre-measured and only need to be diluted with purified water.
Supplier: Avantor
Description: J.T.Baker® Cell Lysis Solution Biotech Reagent is a cGMP pre-formulated and ready to use detergent optimized for the extraction and recovery of AAV pa...
Supplier: Avantor
Description: Volumetric concentrate, for dilution to 500 ml.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 630
no targeter for Bottom