Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Product Category
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "PCR Reagents"
https://ru-bot.vwr.com/stibo/smallweb/std.lang.all/96/19/18989619.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"154"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Peqlab
Description:   peqGOLD Taq DNA Polymerase is a highly processive 5'→3' polymerase with no 3'→5' exonuclease (proofreading) activity. At 72 °C and around pH 9...
Catalog Number: (K461-KIT)
Supplier:  VWR Chemicals
Description:   Set contains one vial of each dNTP (dATP, dTTP, dCTP, dGTP), 40 μmol of 100 mM solution, pH 7,0.
UOM:  1 * 1 KIT
Supplier:  VWR Chemicals
Description:   Extender™ PCR-to-Gel Master Mix, 2X offers a single-step procedure for performing PCR reactions followed by analysis on agarose gels without the addit...
Catalog Number: (N557-0.5ML)
Supplier:  VWR Chemicals
Description:   Ready to use dNTP mixture, 10 mM solution.
UOM:  1 * 0,5 ml
Supplier:  VWR Chemicals
Description:   Ready PCR Mix, 2X offers a single-step procedure for performing PCR reactions followed by analysis on agarose gels without the addition of loading buf...
Supplier:  VWR Chemicals
Description:   Extender™ Taq DNA Polymerase is a high fidelity, thermostable enzyme blend optimised to generate high product yields and purity. This polymerase blend...
UOM:  1 * 1 KIT
Catalog Number: (N224-500U)
Supplier:  VWR Chemicals
Description:   Taq DNA Polymerase is ideal for general PCR and primer extension reactions to amplify DNA templates with high sensitivity. PCR can be initiated from a...
UOM:  1 * 500 items
Supplier:  VWR Peqlab
Description:   A blend of peqGOLD 'Standard' Taq and an inert red dye enables quick PCR set-up and avoids the need for additional dyes for electrophoresis.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 154
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next