You Searched For: Grounders
23  results were found
SearchResultCount:"23"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (115-3067)
Supplier: DESCO
Description: The Dual-Wire thermoplastic wrist strap consists of an insulative exterior with a conductive interior and a 3,4 mm dual conductor jack. The wrist stra...
UOM: 1 * 1 items
Catalog Number: (115-3036)
Supplier: DESCO
Description: MagSnap 360â„¢ dual-wire metal wristband creates a superior electrical connection to eliminate intermittent failures.
UOM: 1 * 1 items
Catalog Number: (115-3047)
Supplier: DESCO
Description: Daisy-chains two worksurface mats for the monitoring of two workstations with the 724 workstation monitor.
UOM: 1 * 1 items
Catalog Number: (115-3045)
Supplier: DESCO
Description: The Desco Europe Slim Earth Bonding Point Bar (or Common Ground Point) has been designed to be installed underneath bench tops where it will be access...
UOM: 1 * 1 items
Catalog Number: (115-3037)
Supplier: DESCO
Description: The MagSnap® 360™ dual wire coil cord offers a 360° rotation and magnetic connection between the wristband and coil cord.
UOM: 1 * 1 items
Supplier: S.C.A.T.
Description: In order to prevent static discharge with fluids in plastic containers, the contents of the waste receptacles can be secured with ground connection. T...
Supplier: Metro International
Description: These grounding cables provide an effective method of electrostatic discharge for any Metro Super Erecta unit (stationary or mobile) when used in conj...
Supplier: Bohlender
Description: For earth connection of vessels and components made of static dissipative materials.
Catalog Number: (115-3055)
Supplier: DESCO
Description: Wrist strap (2209) is designed for one-time use. The wrist strap is ideally suited for shipment along with ESD sensitive items [ESDS], providing the m...
UOM: 1 * 1 items
Catalog Number: (115-3044)
Supplier: DESCO
Description: The EBP bar should be fixed to the edge of a bench or other suitable work equipment within reach and accessible to all connecting ground cords, but wh...
UOM: 1 * 1 items
Catalog Number: (115-3049)
Supplier: DESCO
Description: These grounders will not mark shoes or floors - reducing maintenance costs of touching up on floors. Size adjustable material expands to fit most shoe...
UOM: 1 * 1 items
Catalog Number: (115-3068)
Supplier: DESCO
Description: High visibility bright colour allows supervisor to see that worker is wearing wrist strap.
UOM: 1 * 1 items
Supplier: DESCO
Description: The Dual-wire fabric wristband is available in one size that adjusts to almost any size wrist for reliable protection.
Supplier: DESCO
Description: Coil cord with 2,5 mm polyurethane insulation, conductor is high strength 7-end duplex tinsel for strength and flexibility.
Catalog Number: (115-3046)
Supplier: DESCO
Description: This provides a storage terminal for the wrist cord when the workstation is unoccupied.
UOM: 1 * 1 items
Catalog Number: (115-3056)
Supplier: DESCO
Description: The Universal snap mounting kit consists of 10 mm stud and socket, threaded backing washer, conical nut, long and short screw, ESD common ground point...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 23
no targeter for Bottom