You Searched For: Ergonomic Supports
17  results were found
SearchResultCount:"17"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (BIMO9450-217)
Supplier: bimos
Description: Industrial footrest with clamp lever and non slip rubber tread.
UOM: 1 * 1 items
Catalog Number: (BHIT723103)
Supplier: Sartorius
Description: This elbow pad helps users to feel more comfortable while pipetting. The visco elastic material of the pad relieves contact stress, pain and discomfor...
UOM: 1 * 1 items
Supplier: Portwest
Description: This high performance belt made from 62% polyester and 38% latex offers full support to the wearer. It reinforces the spine, supports the abdomen and ...
Catalog Number: (PRTWKP30BKR)
Supplier: Portwest
Description: These lightweight knee pads consist of a PVC shell with EVA foam and gel filling.
UOM: 1 * 1 PAIR
Catalog Number: (PRTWKP50WHR)
Supplier: Portwest
Description: This knee pad made of EPE foam, polyester provides excellent protection and stability on all floor types.
UOM: 1 * 1 PAIR
Catalog Number: (SIOEYMB00000C0)
Supplier: SIOEN PERSONAL PROTECTIVE CLOT
Description: These knee protectors are recommended for work in the kneeling position. The protection distributes forces evenly and prevents small hard objects on t...
UOM: 1 * 1 items
Catalog Number: (113-8801)
Supplier: Portwest
Description: These durable knee pads are made of ethylene-vinyl acetate. They provide protection against impact injury during, e.g., a fall or a strike, or padding...
UOM: 1 * 1 PAIR
Catalog Number: (PRTWKP40BKR)
Supplier: Portwest
Description: This strap-on gel knee pad offers tough protection for gruelling deep knee work. The thick gel adapts to the wearers knee, providing unrivalled comfor...
UOM: 1 * 1 PAIR
Catalog Number: (PRTWKP05BKR)
Supplier: Portwest
Description: This type 3 kneeling mat is a 'quick fix' knee protection solution for those jobs that simply just will not wait. It is also incredibly useful to have...
UOM: 1 * 1 items
Catalog Number: (PRTWKP20BKR)
Supplier: Portwest
Description: Lightweight knee pads with a tough nylon exterior. The innovative EVA foam centre cleverly moulds to the body shape for complete comfort and protectio...
UOM: 1 * 1 PAIR
Supplier: CAPITAL SAFETY DEUTSCHLAND
Description: Quick Spins simply slide onto the handle of a tool in seconds and provide support.
Catalog Number: (139-9922)
Supplier: WEDO WERNER DORSCH GMBH
Description: Made from steel tubing, with nickel-plated finish.
UOM: 1 * 1 items
Catalog Number: (139-9805)
Supplier: WEDO WERNER DORSCH GMBH
Description: A stylish footrest that is fully adjustable from a normal seated position. Ideal for use at shared workstations.
UOM: 1 * 1 items
Catalog Number: (139-9923)
Supplier: WEDO WERNER DORSCH GMBH
Description: Footrest, relax, PS, Light grey, W×D×H: 490×350×190 mm, Weight: 2,4 kg
UOM: 1 * 1 items
Catalog Number: (KWIN100713930)
Supplier: FRISTADS KANSAS GROUP
Description: Long knee pads made of black foam.
UOM: 1 * 1 items
Supplier: FRISTADS KANSAS GROUP
Description: For anyone who needs to work on their knees.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 17
no targeter for Bottom