You Searched For: Coolers
12  results were found
SearchResultCount:"12"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (AGLS401349)
Supplier: AGILENT
Description: The StrataCooler LP benchtop cooler provides the most protection for enzymes whether in freezer or on benchtop.
UOM: 1 * 1 items
Catalog Number: (AGLS400005)
Supplier: AGILENT
Description: Most cell types show >50% survival when cooling rates are kept at 0,2 to 1 °C/min. The StrataCooler Cryo preservation module has been designed to ...
UOM: 1 * 1 items
Supplier: Cole Parmer
Description: High capacity storage for transport of dry ice pellets and blocks.
Supplier: Corning
Description: Ideal for transfering samples between labs, or in the field.
Supplier: Corning
Description: An open-platform ice-free cooler that accommodates most CoolRack® and CoolSink® modules.
Catalog Number: (SIMPM479-24)
Supplier: Simport Scientific
Description: The Simport KoolPlateâ„¢ is ideal for chilling and preparing paraffin blocks, for sectioning at the microtome or for performing other laboratory procedu...
UOM: 1 * 6 items
New Product
Supplier: HEATHROW
Description: These reagent reservoir chillers are the ice-free, non-toxic way to extend the working period and preserve integrity for temperature sensitive reagent...
Sale
Supplier: GRANT INSTRUMENTS LTD.
Description: Refrigerated coolers are primarily used for counter-cooling when used with a heated circulating bath or for rapidly cooling fluids down to low tempera...
Supplier: DELTA T
Description: Molecular medical elements for different temperature handling of samples ranges from −32 °C to +38 °C.
Catalog Number: (LEIC14039353599)
Supplier: LEICA MICROSYSTEMS
Description: HistoCore Arcadia C is a cold plate holding more than 60/65 cassettes on its large working surface. Cooling efficiency is important, so the cold plate...
UOM: 1 * 1 items
Supplier: CELLPATH
Description: These cooling plate are kept in a freezer or cryostat and placed in the tray to cool tissue blocks. The plate can be cooled to –20 Â°C and keeps t...
Catalog Number: (FEAT501003)
Supplier: PFM Medical GmbH
Description: This cooling plate allows fast and easy cooling of paraffin blocks. The housing is made from stainless steel and the cooling surface is made of anodis...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 12 of 12
no targeter for Bottom