You Searched For: Cleaning Agents
266  results were found
SearchResultCount:"266"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (630-2081)
Supplier: VWR Collection
Description: This cleaning set includes:
- Lens liquid
- Cotton sticks (×5)
- Anti-static cloth (18×15 cm)
- Smart wipes (×2)
- Air pump
UOM: 1 * 1 items
MSDS
Supplier: VWR Chemicals
Description: Compact HDPE canisters (Cat. Nos. 845xx.365) are compatible with Miele dosing cabinets.
Supplier: VWR Chemicals
Description: A range of cleaning agents designed for use in ultrasonic baths.
MSDS Sale
Supplier: VWR Chemicals
Description: Iso-propyl alcohol Ph. Eur. 70% v/v, with purified water Ph. Eur. 30% v/v, filtered at 0,2 μm.
Catalog Number: (88289.360)
Supplier: VWR Chemicals
Description: Alkaline, powerful universal cleaning agent for use in ultrasonic baths or for manual cleaning in an immersion bath.
UOM: 1 * 5 kg
MSDS
Catalog Number: (3900.)
Supplier: Avantor
Description: Decontamination Solution, ProClean cleaner
UOM: 1 * 5 L
MSDS
Supplier: VWR Chemicals
Description: A ready to use, liquid product designed to stabilise and protect water baths, cooling circuits, water circuits and incubators.
Supplier: VWR Chemicals
Description: DNase and RNase contamination ruins expensive samples and time-consuming protocols in research laboratories every day. NucleasEliminatorâ„¢ can help pro...
Catalog Number: (3451.1500)
Supplier: Avantor
Description: Cleaning application: Wax repellent, Paraffin cleaner
UOM: 1 * 1.500 mL
MSDS
Catalog Number: (3768.1000)
Supplier: Avantor
Description: Decontamination Solution, Proclean, micros bottle
UOM: 1 * 1 L
Catalog Number: (94006.1016)
Supplier: VWR Chemicals
Description: Iso-propyl alcohol Ph. Eur. 70% V/V, with purified water Ph. Eur. 30% V/V, filtered at 0,2 μm.
UOM: 1 * 6 L
Catalog Number: (7348.5000)
Supplier: Avantor
Description: RBS 35 concentrate is used for manual cleaning, soaking and ultrasonic baths. These surfactants are alkaline, foaming and insensitive to water hardnes...
UOM: 1 * 5 L
Catalog Number: (00699030.)
Supplier: VWR Chemicals
Description: Acidic neutraliser and pre-cleaner based on organic acid.
UOM: 1 * 5 kg
Catalog Number: (2157-04)
Supplier: MACRON (A AVANTOR BRAND)
Description: CHEM-SOLV® is a non chromic acid cleaner suitable for removing silicone greases, dried blood, and other organic residues from laboratory glass and pla...
UOM: 1 * 1 items
Supplier: Diversey
Description: Concentrated heavy-duty degreaser for cleaning of all heavily soiled surfaces in kitchens like counter tops, stove top and kitchen appliance surfaces....
Supplier: CONTEC
Description: Blend of hydrogen peroxide and peracetic acid.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 266
no targeter for Bottom