You Searched For: Beads
74  results were found
SearchResultCount:"74"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Grinding ball, Stainless steel, Ø: 20 mm
Supplier: VWR Collection
Description: Lightweight, economic PP spheres which form a blanket on the liquid surface. Spheres have 20 mm Ø, 275 spheres cover approximately 1000 cm².
Supplier: VWR Chemicals
Description: Soda-lime glass, solid.
Certificates New Product
Supplier: VWR Collection
Description: Soda-lime glass, solid.
Supplier: VWR Collection
Description: Grinding ball, Set of 5 pieces, Zirconium oxide
Catalog Number: (332124G)
Supplier: VWR Chemicals
Description: Chemically resistant glass balls suitable for use as mixing beads, boiling stones, or packing for distillation columns.
UOM: 1 * 500 g
Catalog Number: (432-0064)
Supplier: VWR Collection
Description: Small beads suitable for use with vortex mixers and tube homogenisers.
UOM: 1 * 200 g
Waste Reducing
Supplier: Thermo Scientific
Description: Pure Al₂O₃ Aluminium oxide, AL 23.
Supplier: Thermo Scientific
Description: Glass beads in glass bottle, Ø: 5,0 mm
Catalog Number: (33190.A1)
Supplier: Thermo Scientific
Description: Grinding pieces, Al-23, 10 mm, 440 pieces/kg, Ceramic
UOM: 1 * 1 kg
MSDS
Catalog Number: (462-8239)
Supplier: GRANT INSTRUMENTS LTD.
Description: Lightweight, economic PP spheres which form a blanket on the liquid surface. Spheres have 20 mm Ø, 275 spheres cover approximately 1000 cm².
UOM: 1 * 300 items
Supplier: Bohlender
Description: Made from solid PTFE, with smooth surface.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Solid soda-lime glass.
Supplier: Thermo Scientific
Description: The density of the zirconium oxide beads is 6.02 g/cm³
Catalog Number: (ROCKTMS-15-50)
Supplier: Rockland Immunochemicals
Description: 6x1,5 ml Position magnetic separator
UOM: 1 * 1 items
Supplier: Thermo Fisher Scientific
Description: The Pierceâ„¢ NHS-activated magnetic beads enable covalent, amine-based conjugation of proteins to magnetic beads in a simple mix-and-go format for use ...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 74
no targeter for Bottom