You Searched For: Animal Supplies
21  results were found
SearchResultCount:"21"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Bel-Art Products, a Part of SP
Description: These universal transparent animal restrainers have removable head and tail gates as well as inner floor.
Supplier: Thermo Scientific
Description: Yeast-Torula
Supplier: MP Biomedicals
Description: Vanderzant vitamin mixture for insects is an animal-derived component free vitamin mixture.
Supplier: MP Biomedicals
Description: This mineral mix, part of AIN-93G rodent diet, is used for growth, pregnancy and lactational phases of rodents.
Supplier: MP Biomedicals
Description: Corn is used as pet nutrition and natural pet food.
Supplier: SOCOREX ISBA
Description: These straight or bent drenchers are made from stainless steel. They are ideal for safe animal nose and mouth feeding.
Supplier: MP Biomedicals
Description: MP recommends the use of supplement U.S.P. XIII, No. 2 Mineral Mix (cat # 960300) in conjunction with this formulation for long term growth and develo...
Catalog Number: (BELAH464200002)
Supplier: Bel-Art Products, a Part of SP
Description: Inhalation narcosis chamber with hinged cover.
UOM: 1 * 1 items
Supplier: MP Biomedicals
Description: AIN-93M powder of proteins, enzymes and peptides is part of the rodent diet.
Catalog Number: (ICNA0290465401)
Supplier: MP Biomedicals
Description: Vitamin diet fortification mixture formulation provides sufficient vitamin levels to prevent deficiencies in all animal species, giving precise contro...
UOM: 1 * 1 kg
Supplier: MP Biomedicals
Description: AIN-76 mineral mixtures are components of an animal diet.
Supplier: MP Biomedicals
Description: An extremely fine, white, odorless tasteless, water insoluble, chemically pure cellulose powder, manufactured from natural fibres.
Catalog Number: (ICNA0290545401)
Supplier: MP Biomedicals
Description: AIN-76 vitamin mixture is a basic component of animal diet for mice and rats colonies in the research lab.
UOM: 1 * 1 kg
Catalog Number: (ICNA0210141301)
Supplier: MP Biomedicals
Description: Corn-syrup is a dual acid-enzyme conversion syrup which contains approximately three times as much maltose as regular corn syrups processed to the sam...
UOM: 1 * 3,79 L
Catalog Number: (ICNA0290283410)
Supplier: MP Biomedicals
Description: This powdered salt mixture is used as animal diet.
UOM: 1 * 10 kg
Catalog Number: (ICNA0296009801)
Supplier: MP Biomedicals
Description: Basic component of animal diet especially for maintaining mice and rats colonies in the research laboratory.
UOM: 1 * 1 kg
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 21
no targeter for Bottom