Palladium(II) nitrate in nitric acid 10%, ARISTAR® matrix modifier solution, for graphite furnace AAS

Supplier: VWR Chemicals
Danger

85769.154EA 0 EUR
85769.154
Palladium(II) nitrate in nitric acid 10%, ARISTAR® matrix modifier solution, for graphite furnace AAS
Palladium(II) nitrate
Formula: N₂O₆Pd
MW: 230,43 g/mol
MDL Number: MFCD00011169
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

Assay (Palladium) 9.8 – 10.2 g/l
Ag (Silver) ≤ 0.1 µg/ml
As (Arsenic) ≤ 0.02 µg/ml
Cd (Cadmium) ≤ 0.005 µg/ml
Pb (Lead) ≤ 0.01 µg/ml
Se (Selenium) ≤ 0.1 µg/ml
Tl (Thalium) ≤ 0.02 µg/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR