VWR® Professional Hotplates
Supplier: VWR
VWR®
444-0628EA 1140 EUR
444-0628 444-0617 444-0627 444-0619 444-0618 444-0624 444-0623 444-0626 VWRU97042-692 444-0625 444-0620 444-0622 444-0621 VWRU97042-732
VWR® Professional Hotplates
Heaters Hotplates
VWR® Professional hotplates are designed for applications that require exceptional accuracy, stability and repeatability. Enhanced microprocessor control offers an external resistance thermometer (RTD) probe option that delivers superior temperature control of the sample. Touch pad controls with easy to read, independent LED displays for temperature and time, allow the operator to view all settings at once. Rear housing features an off-centre, built-in support rod holder with locking knob that accepts the supplied probe kit. Low profile design takes up less space and fits into fume hoods. Spill resistant housing channels fluids away from internal components. A 'hot' symbol warning light is illuminated when heat is turned on and remains on until the plate cools down. Plate over-temperature limit ensures plate temperature will never exceed users' programmed set temperature limit, allowing for control of sensitive flash points. An audible alarm will sound when time reaches zero or when unit reaches set point temperature in timed mode, and if the probe disengages from sample, unit will automatically shut off the heater. Enhanced electronics regulate heating and bring samples to temperature quickly and efficiently. Ceramic tops feature a chemically resistant, reflective white top plate surface that's easy to clean. Durable aluminium tops will not crack or chip, and will provide a more even heating surface.
  • Excellent temperature uniformity
  • Separate digital displays for temperature and time with indicators that toggle between actual and set point. Display will show last used settings, even after power has been turned off
  • Cool touch, chemically resistant housing
  • Includes external RTD temperature probe kit
  • Choice of models with ceramic or aluminium top plates
Delivery information: Each unit is supplied complete with a 203 mm stainless steel PT1000 temperature probe, 457 mm stainless steel support rod, thermometer/temperature probe extension clamp and a hook connector.
Caution: * RT= Ambient